missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Sperm-associated antigen 7 His Protein

Product Code. 18119471 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
This item is not returnable. View return policy

Product Code. 18119471

Brand: Novus Biologicals™ NBP222817

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-227 of Human Sperm-associated antigen 7 The Recombinant Human Sperm-associated antigen 7 Protein is derived from E. coli. The Recombinant Human Sperm-associated antigen 7 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_004881
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 7.5), 0.15 M NaCl, 30% glycerol, 1mM DTT
Gene ID (Entrez) 9552
Molecular Weight (g/mol) 28kDa
Name Sperm-associated antigen 7 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen SPAG7, 1-227aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMADLLGS ILSSMEKPPS LGDQETRRKA REQAARLKKL QEQEKQQKVE FRKRMEKEVS DFIQDSGQIK KKFQPMNKIE RSILHDVVEV AGLTSFSFGE DDDCRYVMIF KKEFAPSDEE LDSYRRGEEW DPQKAEEKRK LKELAQRQEE EAAQQGPVVV SPASDYKDKY SHLIGKGAAK DAAHMLQANK TYGCVPVANK RDTRSIEEAM NEIRAKKRLR QSGEELPPTS
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.