missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PMM2/Phosphomannomutase 2 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.1mg
0.5mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-246 of Human PMM2/Phosphomannomutase 2 The Recombinant Human PMM2/Phosphomannomutase 2 Protein is derived from E. coli. The Recombinant Human PMM2/Phosphomannomutase 2 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 1mg/mL |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol 1mM DTT, 0.1M NaCl. |
| Gene ID (Entrez) | 5373 |
| Molecular Weight (g/mol) | 30.2kDa |
| Purification Method | Protein |
| Quantity | 0.1 mg |
| Source | Human |
| Immunogen | PMM2, 1-246 aa. Sequence: MGSSHHHHHHSSGLVPRGSHMAAPGPALCLFDVDGTLTAPRQKITKEMDDFLQKLRQKIKIGVVGGSDFEKVQEQLGNDVVEKYDYVFPENGLVAYKDGKLLCRQNIQSHLGEALIQDLINYCLSYIAKIKLPKKRGTFIEFRNGMLNVSPIGRSCSQEERIEFYELDKKENIRQKFVADLRKEFAGKGLTFSIGGQISFDVFPDGWDKRYCLRHVENDGYKTIYFFGDKTMPGGNDHEIFTDPRTMGYSVTAPEDTRRICELLFS |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction