missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Quantity | 1 Kit |
| Target | QuikChIP (with Loading Control) |
| Kit Contents | Wash Buffer A 25ml, Wash Buffer B 25ml, Wash Buffer C 25ml, Wash Buffer D 25ml, SDS Lysis Buffer 50ml, 10X Glycine 25ml, 10X PBS 55ml, ChIP Dilution Buffer 50ml, Salmon Sperm DNA/ Protein A/G Agarose 2 x 1.75mL, RNA Pol II Antibody 1mg/ml 25μL, RNAse A 10 mg/ml 60μL, Human GAPDH Primers 10 pmole/μL 100μL, Negative Control Primers 10 pmole/μL 100μL, 100X PMSF 0.75ml, 100X PIC 0.75mL, Proteinase K (10 mg/ml) 60μL, 20% SDS 625μL, 1M NaHCO3 1.25ml, 0.5M EDTA 250μL, 5M NaCl 500μL, 1M Tris-HCl pH 6.5 500μL |
| For Use With (Application) | Western Blotting |
| Product Type | Kit (ChIP) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?