Learn More
Tuttnauer Nozzle Cone
Product Code. 17888289 Shop All Tuttnauer Products
Click to view available options
:
Nozzle Cone
Product Code. 17888289
Brand: Tuttnauer D00543600
Only null left
Add to basket
Add to basket
Plastic cone diameter 15 to 32 mm for nozzles diameter 2.5 mm, nozzle cross diameter 4.
  • Plastic cone diameter 15 to 32 mm for nozzles diameter 2.5 mm.
Specifications
Material Plastic
Product Type Nozzle Cone
For Use With (Equipment) Nozzles (2.5 mm dia.)
Diameter (Metric) 15 to 32 mm