Learn More
Lovibond™ NSB Color of Water Disc
Lovibond NSB Color of Water Disc, Range: 70 to 250 Color Units
Brand:  Lovibond™ 284120
Additional Details : Weight : 0.80000kg
Product Code. 12556279
  • 278.00€ / Each
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
Lovibond NSB Color of Water Disc, Range: 70 to 250 Color Units
Specifications
Specifications
70 to 250 Color Units
Water Disc