missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals NSC 663284

Product Code. 15417919
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 15417919

Brand: Thermo Scientific Chemicals J66520.M^

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 383907-43-5
Molecular Formula C15H16ClN3O3
Synonym 6-Chloro-7-(2-morpholin-4-yl-ethylamino)quinoline-5,8-dione; Cdc25 Phosphatase Inhibitor II

Specifications

Quantity 1 mg
Description Red
Content And Storage -30°C to -10°C
Formula Weight 321.76g/mol
Sensitivity Light Sensitive
Form Crystalline solid
InChI Key BMKPVDQDJQWBPD-UHFFFAOYSA-N
SMILES ClC1=C(NCCN2CCOCC2)C(=O)C2=C(C=CC=N2)C1=O
IUPAC Name 6-chloro-7-{[2-(morpholin-4-yl)ethyl]amino}-5,8-dihydroquinoline-5,8-dione
Molecular Weight (g/mol) 321.76

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.