Learn More
Abnova™ NTRK1/CEN1p FISH Probe
Product Code. 16033325
Change view
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
Product Code. Quantity unitSize
16033325 100 μL 100µL
16043325 200 μL 200µL
2 options
Product Code. 16033325
Brand: Abnova™ FG0202.100uL
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Specifications
Content And Storage Store at 4°C in the dark
Format Tube
For Use With (Application) Fluorescent In Situ Hybridization (Cell)
Gene neurotrophic tyrosine kinase, receptor, type 1
Gene Alias DKFZp781I14186,MTC,TRK,TRK1,TRKA,p140-TrkA
Gene Symbol NTRK1
Gene ID (Entrez) 4914
Product Type DNA/RNA
Quantity 100 μL
Form Liquid