Learn More
Macherey-Nagel™ NucleoBond™ PC EF Kit Buffer Solutions
Product Code. 15889107
Click to view available options
Quantity:
600 mL
1000 mL
Chemical Name or Material:
Acetate buffer solution
Elution Buffer N5-EF
Equilibration Buffer N2-EF
Lysis Buffer S2-EF
Resuspension Buffer S1-EF
Wash Buffer N3-EF
Wash Buffer N4-EF
Water
Unit Size:
1 L
600mL
Product Code. 15889107
Brand: Macherey-Nagel™ 740792.1
Additional supply of buffer for use with NucleoBond™ PC EF kits (PC 500 EF, PC 2000 EF, PC 10000 EF, PC Prep 100) for the purification of endotoxin-free plasmid DNA. Buffer solutions include: S1/S2/S3-EF, N2/N3/N4/N5-EF, TE-EF, and H2O-EF.
NucleBond™ PC EF: The classic anion exchanger for purification of endotoxin-free plasmid DNA
  • Proven anion-exchange technology – transfection-grade plasmid DNA
  • Plasmid DNA with less than 0.1 EU/μg for transfection of highly sensitive cells
  • Patented endotoxin removal by additional washing step – no extra ice incubation necessary
  • Fast and easy clarification of lysates by Folded Filters – no centrifugation required, no shearing forces
  • NucleoBond™ Finalizers available to avoid time consuming centrifugation for plasmid precipitation
  • Separate binding columns of NucleoBond™ PC kits available as NucleoBond™ AX columns
Specifications
Chemical Name or Material Acetate buffer solution
CAS 64-17-5d
Color Colorless
Concentration Acetic acid; Potassium; Sodium + Water
For Use With (Application) Purification of endotoxin-free plasmid DNA
For Use With (Equipment) NucleoBond™ PC EF kits (PC 500 EF, PC 2000 EF, PC 10000 EF, PC Prep 100)
pH 5.2 to 5.7
Physical Form Liquid
Quantity 1000 mL
Formulation Acetic acid; Potassium; Sodium + Water
Odor Acetic