missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ Xtra BAC Buffer Set

Product Code. 12733401
Click to view available options
:
Includes RES-BAC (1000 mL), LYS-BAC (1000 mL), NEU-BAC (1000 mL), RNase A (0.6-100 mg)
This item is not returnable. View return policy

Product Code. 12733401

Brand: Macherey-Nagel™ 740437

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Buffer set for the isolation of large constructs, like BACs, PACs, and P1; only applicable with NucleoBond Xtra BAC kits.

NucleoBond™ Xtra BAC — For transfection-grade, large construct plasmid DNA

  • Optimized column design – one prep in approximately 75 min
  • Column filter included – parallel lysate clearing and loading onto the column
  • Optimal silica material – yields up to 150 μg
  • Transfection-grade plasmid DNA using proven anion-exchange technology
  • LyseControl for visualization of a completed lysis / neutralization
  • Typical downstream applications: cloning, PCR, restriction analysis, sequencing, and transfection

Specifications

Color Colorless (LYS-BAC: Blue)
For Use With (Application) Purification of transfection grade, large construct plasmid DNA
For Use With (Equipment) NucleoBond Xtra BAC Kit
Physical Form Liquid
Quantity 1 Set
Formulation LYS-BAC: Sodium hydroxide solution, diluted; Dodecyl sulfate, sodium salt; Indicator dye
NEU-BAC: Acetate buffer solution
RES-BAC: Chemical mixture solution
RNase A: RNase, lyophized
Odor Odorless (NEU-BAC: Acetic)
Includes RES-BAC, LYS-BAC, NEU-BAC (1000 mL each); RNase A (100 mg)
compliance-icons compliance-icons
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Respiratory sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.