Learn More
Macherey-Nagel™ Nucleogel™ SUGAR 810 Ca HPLC Guard Columns
Brand: Macherey-Nagel™ 719571
Additional Details : Weight : 1.00000kg
Specifications
30 mm | |
4 mm | |
Valco HPLC Guard Column | |
HPLC |
HPLC Guard Column | |
NUCLEOGEL™ SUGAR 810 Ca | |
Separation of Mono, Di and Oligosaccharides | |
L19 |