missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag Tissue Prefilled Plates for DNA Purification from Cells and Tissues

Product Code. 30694918
Change view
Click to view available options
Isolation Technology:
Beads
Unit Size:
96 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Isolation Technology unitSize
30694918 Beads 96 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30694918 Supplier Macherey-Nagel™ Supplier No. 744302

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

NucleoMag Tissue Prefilled Plates offer a ready-to-use solution for purification of DNA from cells and tissues.

  • The reliable chemistry of our NucleoMag Tissue kit is prefilled into convenient deep-well plates.
  • NucleoMag Tissue Prefilled Plates are compatible with the MN extraction robots MagnetaPure 32 Plus and IsoPure Mini.
  • < 20 mg Human/Animal Tissue, < 106 Cells

Specifications

Cell Type Human Tissue, Cells, Animal Tissue
Capacity 0.4 μg/μL Beads
Content And Storage 96 Preps
Format Beads
Isolation Technology Beads
Sample Type Human Tissue,Cells,Animal Tissue
Elution Volume 100 μL
Final Product Type DNA
For Use With (Application) Isolation of DNA
No. of Reactions 96 Prep
Product Line NucleoMag
Product Type Prefilled Plates
Quantity 96 Preps
Purity 1.6 to 1.9
Technique Magnetic separation
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.