Learn More
Macherey-Nagel™ Nucleosil™ 300-5 Standard C18 HPLC CC Guard Columns
Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric) Inner:
4 mm
Unit Size:
Pack of 3
Specifications
Specifications
| Carbon Load | 6.5% |
| pH | 2 to 8 |
| Particle Size | 5 μm |
| Length (Metric) | 8 mm |
| Pore Size | 300Å |
| Type | HPLC CC Guard Column |
| Diameter (Metric) Inner | 4 mm |
| Stationary Phase | NUCLEOSIL™ Standard C18 |
| Mobile Phase Compatibility | Acetonitrile/Water |
| Solvents | Acetonitrile/Water |
| Show More |