missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Blood QuickPure, Mini kit for quick DNA purification from blood
Click to view available options
No. of Reactions:
10
50
250
Unit Size:
10 reactions
250 reactions
50 reactions
Description
Features:
- For ultra-fast purification of highly concentrated DNA from blood
- Ultra-fast procedure
- Washing and drying in one combined step (hands-on-time < 10 minutes)
- Consistently high DNA yields
- Elution volume only 30 to 50μL
- Highly concentrated DNA, ready to use for sensitive downstream applications
Applications:
- DNA from whole blood (human or animal, fresh or frozen)
- DNA from whole blood treated with citrate, EDTA, heparin, CPDA
- DNA from buffy coat, platelets, body fluids (e.g. amniotic fluid), serum, plasma
- DNA from cultured cells
- Typical downstream applications: PCR, Southern blotting, enzymatic reactions
Specifications
Specifications
| Format | Mini Spin Column |
| Isolation Technology | Silica Membrane Technology |
| For Use With (Application) | Applications: DNA from whole blood (human or animal, fresh or frozen), DNA from whole blood treated with citrate, EDTA, heparin, CPDA, DNA from buffy coat, platelets, body fluids (e.g., amniotic fluid), serum, plasma, DNA from cultured cells, Typical downstream applications: PCR, Southern blotting, enzymatic reactions. |
| No. of Reactions | 250 |
| Product Type | DNA purification column |
| Quantity | 2 mL |
| Test Time | 25 min. |
Safety and Handling
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction