Learn More
Macherey-Nagel™ NucleoSpin™ Tissue, Mini kit for DNA from cells and tissue
Click to view available options
No. of Reactions:
10
12 Preps
250
50
Unit Size:
10 reactions
12 preps
250 reactions
50 reactions
Description
- Versatile kit for the isolation of genomic DNA from a wide range of different starting materials (cells, tissue, forensic and clinical samples)
- 16 support protocols available in the user manual for highest flexibility
- Highly sensitive membrane for reliable purification of even trace amounts of DNA
- Rapid and easy protocol (20 min./prep excl. lysis)
Specifications
Specifications
| Buffer | Yes |
| For Use With (Application) | DNA |
| Includes | Lysis Buffer T1, Buffer B3, Wash Buffer BW, Wash Buffer B5, Elution Buffer BE, Proteinase K, Proteinase Buffer PB, NucleoSpin Blood Columns, User manual |
| No. of Reactions | 12 Preps |
| Product Type | NucleoSpin Tissue Kit |
| Quantity | 12 Pack |
Safety and Handling
Product Identifier
- NucleoSpin Tissue
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : guanidine hydrochloride / 2-propanol