Learn More
Trajan Scientific and Medical™ Nuts for Varian Injector
Gives low dead volume connections for optimum chromatography.
Brand:  Trajan Scientific and Medical™ 1034060
Additional Details : Weight : 0.01000kg
Product Code. 15259113
  • 35.50€ / Pack of 2
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
Description
Description
  • Low dead volume
  • High temperature applications
  • Leak free connections
Specifications
Specifications
Nuts for Varian Injector
2/Pk.