missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NVS-STG2

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
NVS-STG2 is a STING agonist. It induces oligomerization of human STING by binding to pockets between adjacent STING dimer transmembrane domains, acting as a molecular glue. NVS-STG2 elicits potent STING-mediated immune responses in cells and produces antitumor activities in hSTING knock-in mice.
Specifications
Specifications
| Chemical Name or Material | 4-((4-(2-(tert-Butyl)-5,5-dimethyl-1,3-dioxan-2-yl)benzyl)amino)-3-methoxybenzoic acid |
| CAS | 3030588-01-0 |
| Quantity | 10 mg |
| Target | STING Agonists |
| Molecular Formula | C25H33NO5 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction