missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NX 5948

Product Code. 30279894
Click to view available options
:
5 mg
This item is not returnable. View return policy

Product Code. 30279894

Brand: Tocris Bioscience™ 8856/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective BTK Degrader (PROTAC™)

NX 5948 is a potent and selective Bruton's tyrosine kinase (BTK) Degrader (PROTAC™) (DC50 = <1 nM; Dmax = 98%). Comprises a BTK binding moiety joined by a linker to cereblon (CRBN) ligand. In vitro, NX-5948 very potently, rapidly and completely degrades BTK in primary human B cells (DC50 = 0.034 nM). NX 5948 degrades BTK, inhibits tumor growth and increases survival in TMD8 xenograft mouse models of brain maligancy. Reduces inflammation and symptom severity in a mouse model of collagen-induced arthritis. NX 5948 is orally bioavailable and blood brain barrier penetrant.

PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[[4-[1-[[1-[6-[[[(3S)-2,6-Dioxo-3-piperidinyl]amino]carbonyl]-3-pyridinyl]-4-piperidinyl]methyl]-4-piperidinyl]phenyl]amino]-5-[(3R)-3-(3-methyl-2-oxo-1-imidazolidinyl)-1-piperidinyl]-2-pyrazinecarboxamide
CAS 2649400-34-8
Quantity 5 mg
Target Active Degraders
Molecular Formula C42H54N12O5
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.