Learn More
Masterflex™ Nylon Barbed Elbow Connector
Click to view available options
Port Size:
2-port, 1/2 in. I.D. tubing
Unit Size:
Pack of 10
Description
- 90° bend tubing connector
- Barb connector
Specifications
Specifications
| Description | 90° elbow tubing connector |
| Autoclavable | Not Autoclavable |
| Material | Nylon |
| For Use With | Flexible tubing |
| Port Size | 2-port, 1/2 in. I.D. tubing |
| Sterilization Method | EtO only |