Learn More
Masterflex™ Nylon Female Luer Fitting
Product Code. 15101425
Click to view available options
For Use With:
Female Luer x 1/8 in. Wide Bore Hose Barb
Female Luer x 1/8-27 NPT Elbow
Female Luer x 3/32 in. Hose Barb
Female Luer x 3/32 in. Wide Bore Hose Barb
Unit Size:
Pack of 25
Product Code. 15101425
Brand: Masterflex™ 4550203
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Use these low-profile hose barbs with semi-rigid tubing such as nylon, polyethylene and polypropylene. Nylon Female Luer Fittings feature a smooth bore hose barb to reduce flow turbulence and risk of line contamination.
  • Standard hose barb for use with flexible tubing
  • Smooth bore hose barb to reduce flow turbulence and risk of line contamination
  • Wide-bore hose barb for enhanced fluid path flow
Specifications
Material Nylon
For Use With Female Luer x 3/32 in. Wide Bore Hose Barb
Packaging Quantity 25 Pack
Product Type Female Luer Fitting