Learn More
Brady™ Nylon Printer Label
Product Code. p-8001600
Click to view available options
Dimensions (LxW):
100 ft. x 2 in.
12.70 x 12.70 mm
12.70 x 19.05 mm
12.70 x 22.86 mm
25.40 x 25.40 mm
25.40 x 48.26 mm
6.35 x 38.10 mm
9.10 m x 12.70 mm
9.10 m x 48.26 mm
9.53 mm x 12.7 mm
Shape:
Rectangle
Rectangle with Rounded Corners
Square
Unit Size:
Each
Product Code. 12860059
Brand: Brady™ 114862
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Self-adhesive repositionable labels
  • Permanent acrylic adhesive
  • White color
  • For general, industrial, laboratory, and panel identification
Specifications
Description Low profile with high adhesion
Adhesive Type Permanent Acrylic
Color White
Finish Matte
Material Nylon Cloth
Product Type Wire Marker Label
For Use With (Application) For general, industrial, laboratory, and panel identification
Labels per Row 1
Shape Rectangle
Size (Metric) 22.86 x 12.7 mm (W x H)
Dimensions (LxW) 12.70 x 22.86 mm