Learn More
Linker Industrietechnik™ Nylon Sieve Filter, 1m x 1m
For sieving or precision filtration of biological and chemical solutions. Linker Industrietechnik™ Nylon Sieve Filters, 1m x 1m are available in a comprehensive range of pore sizes.
Brand: Linker Industrietechnik™ 1000X1000M600M
Additional Details : Weight : 0.80000kg
Description
- Resistant to aggressive solvents
- Excellent compatibility with alcohols
- Operating temperature of -40° to +115°C
- Applications include solvent filtration and particle analysis
Specifications
Nylon | |
Plastic | |
600μm |