Learn More
Masterflex™ Nylon T Connector
Product Code. 12354628
Click to view available options
For Use With:
4.76mm
6.35mm
9.52mm
12.7mm
19.05mm
25.4mm
Unit Size:
Pack of 10
Product Code. 12354628
Brand: Masterflex™ 3061330
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Diameter (Metric) Inner 19.05 mm
Material Nylon
For Use With 19.05mm
Min. Operating Temperature (Metric) -46°C
Packaging Quantity 10 Pack
Shape T
Max. Operating Temperature (Metric) 121°C
Product Type T-Connector