Learn More
Masterflex™ Nylon Threaded Fittings
Click to view available options
For Use With:
0.125 in. NPT(M)
0.125in.
0.25 in. NPT(M)
0.25in. Tubing
0.375 in. NPT(M)
0.375in.
0.75 x 0.5in.
0.75in.
Unit Size:
Pack of 10
Pack of 6
Specifications
Specifications
| Material | Nylon |
| For Use With | 0.25in. Tubing |
| Packaging Quantity | 6 Pack |
| Product Type | Threaded Fitting |