Learn More
Masterflex™ Straight Fitting, Threaded Nipple, NPT(M) x NPT(M), Nylon
Click to view available options
Thread Size:
3/8 in. NPT
Unit Size:
Pack of 6
Description
- Easy to use
- Secure threaded connection
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Material | Nylon |
| For Use With | 3/8 in. NPT thread |
| Min. Operating Temperature (Metric) | -40°C |
| Packaging Quantity | 6 Pack |
| Sterilization Method | Ethylene Oxide |
| Thread Size | 3/8 in. NPT |
| Maximum Pressure | 200 psi |
| Max. Operating Temperature (Metric) | 93°C |
| Product Type | Threaded Nipple NPT(M) Connector |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur