missing translation for 'onlineSavingsMsg'
Learn More

o-Tolidine dihydrochloride, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-7077328
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
612-82-8
C14H16N2·2HCl
285.2
LUKPNZHXJRJBAN-UHFFFAOYSA-N
3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl
108938
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl
This item is not returnable. View return policy
612-82-8
C14H16N2·2HCl
285.2
LUKPNZHXJRJBAN-UHFFFAOYSA-N
3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl
108938
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 130.0°C
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Quantity 25 g
Merck Index 15,9675
Solubility Information Passes test
Formula Weight 285.2
Grade ACS Reagent
Chemical Name or Material o-Tolidine dihydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'