missing translation for 'onlineSavingsMsg'
Learn More

o-Tolylacetonitrile, 98+%, Thermo Scientific Chemicals

Catalog Number 11432517
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
22364-68-7
C9H9N
131.178
MFCD00001904
WMGVPDQNPUQRND-UHFFFAOYSA-N
2-methylbenzyl cyanide, 2-o-tolyl acetonitrile, o-tolylacetonitrile, 2-methylphenylacetonitrile, o-methylphenylacetonitrile, o-xylyl cyanide, o-methylbenzyl cyanide, acetonitrile, o-tolyl, benzeneacetonitrile, 2-methyl, 2-2-methylphenyl acetonitrile
31155
CHEBI:27982
2-(2-methylphenyl)acetonitrile
CC1=CC=CC=C1CC#N
This item is not returnable. View return policy
22364-68-7
C9H9N
131.178
MFCD00001904
WMGVPDQNPUQRND-UHFFFAOYSA-N
2-methylbenzyl cyanide, 2-o-tolyl acetonitrile, o-tolylacetonitrile, 2-methylphenylacetonitrile, o-methylphenylacetonitrile, o-xylyl cyanide, o-methylbenzyl cyanide, acetonitrile, o-tolyl, benzeneacetonitrile, 2-methyl, 2-2-methylphenyl acetonitrile
31155
CHEBI:27982
2-(2-methylphenyl)acetonitrile
CC1=CC=CC=C1CC#N

o-Tolylacetonitrile is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an organic and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Tolylacetonitrile is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an organic and chemical intermediate.

Solubility
Slightly soluble in water. Soluble in benzene, ethyl ether, ethanol.

Notes
Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances.
TRUSTED_SUSTAINABILITY
Density 1.01
Boiling Point 211°C to 213°C
Flash Point >110°C (230°F)
Odor Almond-like
Refractive Index 1.5275
Quantity 100 g
UN Number UN3276
Beilstein 907182
Solubility Information Slightly soluble in water. Soluble in benzene,ethyl ether,ethanol.
Formula Weight 131.18
Percent Purity ≥98%
Chemical Name or Material o-Tolylacetonitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'