missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation Oasis MCX Online Cartridge Column
The Oasis MCX on-line cartridge column is designed for rugged, reproducible, and ultra-fast on-line SPE.
Brand: Waters Corporation 186002051
Additional Details : Weight : 0.50000kg
Description
- This cartridge column contains the Oasis MCX sorbent, which is a novel, mixed-mode polymeric sorbent that has been optimized to achieve higher selectivity and sensitivity for extracting basic compounds with cation-exchange groups.
- This on-line cartridge column is designed to be used with the appropriate Waters narrow-bore analytical column, such as XSelect, XBridge, SunFire, Atlantis, XTerra, or Symmetry.
- The Oasis MCX on-line cartridge column fits into a Sentry holder that features a finger-tight fitting for fast, convenient replacement.
Specifications
2.1 mm | |
Spherical | |
80 Å | |
No |
20 mm | |
30 μm | |
1/Ea. | |
Cartridge |