Learn More
Carl Zeiss™ Objective EC Plan Neofluar
Use these universal objectives for all contrast methods: bright field, dark field, phase contrast, DIC, fluorescence and Pol. Zeiss™ Objective EC Plan Neofluar feature high resolving power and brilliant, rich-contrast, completely flat images for observation and documentation.
Brand: Carl Zeiss™ 420360-9900-000
Additional Details : Weight : 0.01000kg
Description
- Enhanced Contrast Plan-Neofluar
- Minimized stray light: with a visible increase in contrast important
- High transmission of wavelengths from the near UV range (>40% starting at 340nm)
- Glass types of low autofluorescence makes these objectives
- Ideal for epi-fluorescence microscopy
Specifications
Objective EC | |
M27, N.A. of 0.75, WD of 0.71mm | |
40X | |
1 Ea. |