Learn More
Carl Zeiss™ Objective EC Plan Neofluar
Use these universal objectives for all contrast methods: bright field, dark field, phase contrast, DIC, fluorescence and Pol. Zeiss™ Objective EC Plan Neofluar feature high resolving power and brilliant, rich-contrast, completely flat images for observation and documentation.
2358.50€ - 4934.30€
Specifications
Type Objective EC
Quantity 1 Ea.
View More Specs
Products 3
Product Code Brand Magnification Power Price Quantity & Availability  
Product Code Brand Magnification Power Price Quantity & Availability  
11875560
View Documents Promotion Details
Carl Zeiss™
420350-9900-000
20X
2358.50€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 08-07-2024
to see stock.
Add to basket
11805123
View Documents Promotion Details
Carl Zeiss™
420360-9900-000
40X
2936.20€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 08-07-2024
to see stock.
Add to basket
10772444
View Documents Promotion Details
Carl Zeiss™
420490-9900-000
100X
4934.30€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 08-07-2024
to see stock.
Add to basket
Description
Description
  • Enhanced Contrast Plan-Neofluar
  • Minimized stray light: with a visible increase in contrast important
  • High transmission of wavelengths from the near UV range (>40% starting at 340nm)
  • Glass types of low autofluorescence makes these objectives
  • Ideal for epi-fluorescence microscopy
Specifications
Specifications
Objective EC
1 Ea.