Learn More
Motic™ Objective Lens for SMZ160
The Objective Lens is compatible with the SMZ-160 microscope.
88.00€
Product Code | Brand | Magnification Power | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Magnification Power | Price | Quantity & Availability | |||||
17827285
|
Motic™
1101001708251 |
0.35X |
88.00€
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
17814501
|
Motic™
1101001708261 |
0.5X |
88.00€
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
17817285
|
Motic™
1101001708271 |
0.75X |
88.00€
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
17807295
|
Motic™
1101001708281 |
1.5X |
88.00€
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
17824501
|
Motic™
1101001708291 |
2X |
88.00€
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
Description
- Objective Lens compatible with SMZ160.
- Not compatible with standard stands.
- Available in sizes 0.35x with WD = 287 mm, 0.5x with WD = 165 mm, 0.75x with WD = 120 mm, 1.5x with WD = 47 mm and 2.0 with WD = 30 mm.
Specifications
SMZ160 Objective | |
SMZ-160 |