Learn More
Fisherbrand™ Octahedral PTFE Stir Bar
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
13 mm
15 mm
22 mm
25 mm
41 mm
51 mm
75 mm
Diameter (Metric):
3 mm
8 mm
13 mm
Unit Size:
Pack of 10
Pack of 5
Description
- Stable, general-purpose magnetic stir bar
- The flat surfaces provide a greater mixing action
- All have pivot rings
- AlNiCo-V magnets encased in PTFE
- Autoclavable
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
| Color | White |
| Description | PTFE encapsulated Alnico V magnet core |
| For Use With (Equipment) | Containers & Vessels / Stirrer Hot Plates |
| Coating | PTFE |
| Diameter (Metric) | 8 mm |
| Length (Metric) | 51 mm |
| Quantity | 10/Pk. |
| Shape | Octahedral |
| Show More |