missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oil Red O, Biological Stain

Product Code. p-7203412 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15244664

Brand: Thermo Scientific Chemicals 189400250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1320-06-5 | C26H24N4O | 408.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1320-06-5
Molecular Formula C26H24N4O
Molecular Weight (g/mol) 408.51
MDL Number MFCD00003898
InChI Key HJAYEODIXUYVIC-LTFAAIOASA-N
Synonym C.I. 26125, Solvent Red 27
PubChem CID 6046885
SMILES CC1=CC=C(C)C(=C1)N=NC1=CC(C)=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C=C1C

Specifications

IUPAC Name (1Z)-1-[[4-[(2,5-dimethylphenyl)diazenyl]-2,5-dimethylphenyl]hydrazinylidene]naphthalen-2-one
Formula Weight 408.5
Quantity 25 g
Chemical Name or Material Oil Red O

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.