Learn More
Sartorius™ OIML Class E2 Proof Line Leaf Weight without Certificate
Product Code. p-7112984 Shop All Sartorius Products
Click to view available options
Mass:
1 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
Density:
2,650kg/m3
8,600kg/m3
Material:
Aluminum
Nickel Silver
Unit Size:
Each
Product Code. 11343334
Brand: Sartorius™ YCW0121AC00
Only null left
Add to basket
Add to basket
Constructed from non-magnetizable aluminum or nickel silver. Sartorius™ OIML Class E2 Leaf Weight without Certificate is higly resistant to corrosion.
  • Milligram weights are provided as leaf weights
  • With nominal mass values ranging from 1mg to 5mg are made of aluminum has density of 2,650 kg/m3
  • Weights with nominal values of 10mg to 500mg are made of nickel silver has density of 8,600 kg/m3
  • Tolerances comply with OIML R111-1
TRUSTED_SUSTAINABILITY
Specifications
Product Type Proof Line Weight
Classification OIML Class E2
Material Aluminum
Density 2,650kg/m3
Mass 1 mg