Learn More
Ohaus™ OIML Class E2 Weights
OHAUS™ OIML Class E2 Weights are adjusted to meet or exceed OIML E2 Tolerances.
Brand: Ohaus™ 11117001
Additional Details : Weight : 0.03000kg
Description
- Constructed of stainless steel
- Weights include a weight case
- Single body design without adjusting cavity
Specifications
OIML E2 Weights | |
0.030 kg |