missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ OIML Class F2 Proof Line Weight Set without Certificate
Includes the same features and properties as the individual weights of this series, along with the corresponding maximum permissible errors. Sartorius™ OIML Class F2 Weight Set without Certificate is supplied with gloves, tweezers and brush in a high-quality plastic case
227.00€ - 1580.00€
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
11383354
|
Sartorius™
YCS01-254-AC-00 |
1 mg to 500 mg |
227.00€
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
11343364
|
Sartorius™
YCS31-514-AC-00 |
1 g to 100 g |
445.00€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
11343374
|
Sartorius™
YCS31-614-AC-00 |
1 g to 1 kg |
905.00€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
11323384
|
Sartorius™
YCS31-654-AC-00 |
1 g to 5 kg |
1580.00€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
Description
Values 1 to 5mg: Aluminum leaf weights with density of 2,650 kg/m3Values of 10 to 500mg: Nickel silver construction with density of 8,600 kg/m3.
Values of 1g and higher: Polished, special stainless steel knob weights with density of 7,950kg/m3
- Highly corrosion resistant
- Non-magnetizable
- Tolerances comply with OIML R111-1
- Includes sturdy plastic carry case
Specifications
Proof Line Weight Set | |
OIML Class F2 |