Learn More
Diba Omnifit™ O-Rings for Chromatography Columns
Click to view available options
Diameter (Metric) Inner:
3 mm
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Material:
FFKM
FKM
Unit Size:
Pack of 10
Pack of 2
Pack of 6
Specifications
Specifications
| For Use With (Equipment) | Omnifit™ HiT Chromatography Columns |
| Quantity | 6 Pk. |
| Diameter (Metric) Inner | 25 mm |
| Type | Liquid Chromatography Column |
| Length (Metric) | 25 mm |
| Material | FKM |