Learn More
Diba Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Columns
Product Code. 15845284
Encompass_Preferred
Click to view available options
Diameter (Metric) Inner:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Length (Metric):
50 mm
100 mm
150 mm
250 mm
330 mm
400 mm
500 mm
Unit Size:
Each
Product Code. 15845284
Brand: Diba 006ERG1010
This item requires a price update prior to being ordered.
Request a Price for this item here.
Mix and match your Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Column lengths while using your existing endpieces for ultimate flexibility
Use a different length tube with your existing endpieces to meet your specific requirements.
Specifications
For Use With (Equipment) Omnifit™ EZ and EZ SolventPlus™ Columns
Diameter (Metric) Inner 10 mm
Type Liquid Chromatography Column
Length (Metric) 100 mm
Material Glass