Learn More
Diba Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Columns
Click to view available options
Diameter (Metric) Inner:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Length (Metric):
50 mm
100 mm
150 mm
250 mm
330 mm
400 mm
500 mm
Unit Size:
Each
Description
Use a different length tube with your existing endpieces to meet your specific requirements.
Specifications
Specifications
| For Use With (Equipment) | Omnifit™ EZ and EZ SolventPlus™ Columns |
| Diameter (Metric) Inner | 15 mm |
| Type | Liquid Chromatography Column |
| Length (Metric) | 400 mm |
| Material | Glass |