missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Oncostatin M Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHOSMX10
Additional Details : Weight : 2.50000kg
Description
Oncostatin M is a member of a cytokine family that includes leukemia-inhibitory factor, granulocyte colony-stimulating factor, and interleukin 6. This gene encodes a growth regulator which inhibits the proliferation of a number of tumor cell lines. It regulates cytokine production, including IL-6, G-CSF and GM-CSF from endothelial cells.Specifications
P13725 | |
1 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
OSM | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
1.37 to 1000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
5008 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Plasma, 50 μL|Serum, 50 μL|Supernatant, 100 μL | |
Oncostatin M | |
4 hr 45 min |