missing translation for 'onlineSavingsMsg'
Learn More

Ohaus™ One Block Digital Dry Block Heater HB1DG

Product Code. 15782699 Shop All Ohaus Products
Click to view available options
No. of Blocks:
1
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15782699

Brand: Ohaus™ 30392061

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

OHAUS™ One Block Digital Dry Block Heater processes more samples while delivering precise and reliable results.

  • Designed for applications that require repeatable results and superior temperature stability
  • These multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a wide variety of other laboratory procedures
  • Integral support rod holder with locking knob accepts optional External Temperature Probe Kit
  • External Temperature Probe Kit monitors actual block or sample temperature
  • Each of the five models accepts separate interchangeable modular blocks, accommodating various tube sizes from 0.2 mL microtubes to 50 mL centrifuge tubes
  • Each block has a thermometer well for measuring block temperature
  • Efficient heating due to close tube-and-block contact for maximum heat retention
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance TUV
Controller Type Digital
Description Dry Block Heater, 1 Block, HB1DG, EU
Temperature Range (Metric) Ambient +5° to 120°C
Type Dry Bath Block
Heat Up Time 45 min
Model HB1DG
No. of Blocks 1
Temperature Stability ±0.1°C
Temperature Uniformity ±0.1°C at 37°C
Uniformity ± 0.2°C at 37°C
Dimensions (L x W) 31.50 cm x 20.30 cm
Dimensions (L x W x H) 31.5 x 20.3 x 8.9 cm
Length (English) 12.40 in.
Length (Metric) 31.50 cm
Width (English) 8.00 in.
Width (Metric) 20.30 cm
Height (English) 3.50 in.
Height (Metric) 8.900 cm
Depth (English) 12.40 in.
Depth (Metric) 31.50 cm
Weight (Metric) 2.5 kg
Electrical Requirements 230V, 0.5A, 50/60 Hz
Amperage 0.5A
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.