Learn More
Hamilton™ One-Hub Assembled Tubing Assemblies
Providing a variety of connection solutions in the laboratory.
Brand: Hamilton™ 90614
Additional Details : Weight : 0.01000kg
Description
- Fitting manufactured from CTFE
- Tubing manufactured from PTFE
- For female luers
- A variety of gauges and lengths available
Specifications
CTFE (Fitting), PTFE (Tubing) | |
304.8 mm | |
1/Pk. | |
Female Luer |
14 G | |
Tubing Assembly | |
14 ga. Connections |