Learn More
Merck Optex™ GS MCE Filtration Membrane
Brand:  Merck SPGS0255W
274.00 EUR valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10215603
  • 294.00€ / Pack of 50
Estimated Shipment: 19-06-2024
to see stock.
Specifications
0.22 μm
Sterile
50/Pk.