Learn More
Merck Optex™ GS MCE Filtration Membrane
Brand: Merck SPGS0250P
2200.00 EUR valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.10000kg
Specifications
0.22 μm | |
Sterile | |
1000/Pk. |