Learn More
Tork OptiServe™ Coreless Toilet Paper T7
GREENER_CHOICE
Product Code. p-200113556 Shop All Tork Products
Click to view available options
Color:
White
Ply:
2-ply
Unit Size:
24 rolls
Product Code. 30243795
Brand: Tork 472630
Only null left
Add to basket
Add to basket
Tork OptiServe™ Coreless is a high-capacity toilet paper system for facilities where efficiency, sustainability and providing a great experience are key.
  • Refill less often, reduce paper shortages, and save storage space as this compact coreless toilet paper has 3x more paper and uses half the storage space compared to conventional rolls.
  • Reduce waste with coreless toilet paper, which doesn't have cores or wraps.
  • Choose from a wide range of Tork OptiServe™ Coreless dispensers to meet various washroom needs, so you can use coreless refills across the facility.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Bathroom Tissue
Length (English) 103.13 yd.
Length (Metric) 94.3 m
Width (English) 3.66 in.
Width (Metric) 9.3 cm
Color White
Ply 2-ply