missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ Csp6I (CviQI), Fisher BioReagents™

Product Code. 11934822
Click to view available options
Quantity:
1500 U
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11934822

Brand: Fisher BioReagents BP80681

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

5'...G^T A C...3'
3'...C A T^G...5'

Supplied with: 10X OPTIZYME Buffer 1
Conditions for 100% Activity:

  • 1X OPTIZYME Buffer 1: 10mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl)2 and 0.1mg/ml BSA
  • Incubate at 37°C
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 100%
  • Buffer 2: 50 - 100%
  • Buffer 3: 0 - 20%
  • Buffer 4: 50 - 100%
  • Buffer 5: 0 - 20%

Storage Buffer:
  • 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
  • Ligation and Re-cleavage:
  • More than 95% of the DNA fragments can be ligated and re-cut after 50-fold over-digestion with Csp6I.
  • Compatible Ends: FspBI, NdeI, Tru1I, VspI
  • Methylation Effects:
  • Dam: never overlaps - no effect
  • Dcm: never overlaps - no effect
  • CpG: may overlap - no effect
    • 5'...m5C GTAm5C G...3'
  • 3'... Gm5CAT Gm5C...5'
  • (No effect on cleavage)
  • EcoKI: never overlaps - no effect
  • EcoBI: never overlaps - no effect
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 10 U/μL
    Components 25 to 50% Water, >50% Glycerin
    Incubator Temperature 37°C
    pH 7.4
    For Use With (Application) >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with Csp6I
    Storage Buffer 10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol
    Content And Storage Keep container tightly closed
    Quantity 1500 U
    Cut Site G.TAC
    Product Type Csp6 I
    Form Liquid
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.