missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPTIZYME™ ScaI, Fisher BioReagents™
Brand: Fisher Bioreagents BP8037-1
Additional Details : Weight : 0.01000kg
Description
5'...A G T^A C T...3'
3'...T C A^T G A...5'
Supplied with: 10X OPTIZYME AarI, ScaI, PasI Buffer
Conditions for 100% Activity:
- 1X OPTIZYME Buffer AarI, ScaI, PasI: 10mM Bis-Tris Propane-HCl (pH 6.5 at 37°C), 10mM MgCl2 , 100mM KCl and 0.1mg/ml BSA
- Incubate at 37°C
- Buffer 1: 0 - 20%
- Buffer 2: 0 - 20%
- Buffer 3: 0 - 20%
- Buffer 4: 0 - 20%
- Buffer 5: 0 - 20%
- 10mM Tris-HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.5mg/ml BSA and 50% (v/v) glycerol
- Conditions of low salt, high enzyme concentration, glycerol concentration >5% or pH >8.0 may result in star activity.
- Supercoiled plasmids may require up to 20-fold more ScaI for complete digestion than linear DNA.
Dam: Never overlaps - no effect
Dcm: Never overlaps - no effect
CpG: Never overlaps - no effect
EcoKI: Never overlaps - no effect
EcoBI: May overlap - blocked
- 5'...TGm6AGTACT(N)3 TGCT...3'
Specifications
10 U/μL | |
37°C | |
>80% of DNA fragments can be ligated and >90% of these can be re-cut after 50-fold over-digestion with ScaI | |
1000 U | |
ScaI |
10X OPTIZYME™ AarI, ScaI, PasI Buffer | |
7.4 | |
10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM EDTA, 1mM DTT, 0.2mg/mL BSA, 50% Glycerol | |
AGT.ACT | |
Liquid |