Learn More
STEMCELL Technologies Organoid Embedding Sheet (6-Pack)
Click to view available options
Quantity:
6 Sheets
Unit Size:
Pack of 6
Description
- The Organoid Embedding Sheet is designed to facilitate embedding 3-dimensional (3D) aggregates (e.g. embryoid bodies, spheroids, or organoids) into a matrix
- The wells on the sheet are designed to fit aggregates up to 5 mm in diameter
- The sheet may be grasped at the top, along the label, for easy handling
- The Organoid Embedding Sheet can also be a substitute for Parafilm™ and used to mix small volumes in various protocols
- Each sheet contains 18 wells, and the sheet is compatible with STEMCELL media products such as STEMdiff™ Cerebral Organoid Kit (Catalog #08570)
Specifications
Specifications
| Quantity | 6 Sheets |