Learn More
STEMCELL Technologies Organoid Embedding Sheet (6-Pack)
Product Code. 17199941
Encompass_Preferred
Click to view available options
Quantity:
6 Sheets
Unit Size:
Pack of 6
Product Code. 17199941
Brand: STEMCELL Technologies 08579
Silicone sheet designed to facilitate embedding 3D aggregates in a matrix
  • The Organoid Embedding Sheet is designed to facilitate embedding 3-dimensional (3D) aggregates (e.g. embryoid bodies, spheroids, or organoids) into a matrix
  • The wells on the sheet are designed to fit aggregates up to 5 mm in diameter
  • The sheet may be grasped at the top, along the label, for easy handling
  • The Organoid Embedding Sheet can also be a substitute for Parafilm™ and used to mix small volumes in various protocols
  • Each sheet contains 18 wells, and the sheet is compatible with STEMCELL media products such as STEMdiff™ Cerebral Organoid Kit (Catalog #08570)
Specifications
Quantity 6 Sheets