Learn More
Thermo Scientific™ Orion™ AQUAfast™ AQ3170 Colorimeter
Gather accurate chlorine measurements with a colorimeter designed for both the laboratory and the field in markets such as drinking water, wastewater, food & beverage, aquaculture, and agriculture.
Brand: Thermo Scientific™ AC2V10
Additional Details : Weight : 0.02600kg
Description
The Thermo Scientific™ Orion™ AQUAfast™ AQ3170 colorimeter has a rugged and waterproof design to provide accurate chlorine measurements whether testing in the laboratory or the field. This colorimeter is designed to improve the user experience by including a large, backlit display, a real time clock and automatic data logging. The AQUAfast AQ3170 colorimeter is used in markets such as drinking water, wastewater, food & beverage, aquaculture, and agriculture.
- Large digital backlit display
- Automatic switch-off
- User friendly keypad
- Real time clock and date
- Calibration mode
- Storage function
- Waterproof sample chamber
- Angled, ergonomic design for easy bench top or hand-held use
- Suppression of chloride interference
- Ability to export data to a PC
- ST-COD tubes ready for use
- 2-year warranty
Specifications
AQUAfast 10mm short path-length plastic vials for high level chlorine, 12 pack | |
AQ3170 Chlorine Colorimeter | |
12 pack of 10 mm plastic short path-length vials |