Learn More
Thermo Scientific™ Orion™ Electrode Adapter Cables
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
Brand:  Thermo Scientific™ 090034
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11820491
  • 116.00€ / Each
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
Description
Description
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
  • For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2mm reference pins or just a stripped lead inputs
  • Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors
Specifications
Specifications
BNC
Meters That Have BNC
E DIN (Knick, Schott, WTW)
E DIN (Knick, Schott, WTW); BNC; BNC to E DIN Adapter
Electrode Cable
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Pierce Iodoacetamide, No-Weigh Format
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P264-Wash hands thoroughly after handling.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor / physician
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P363-Wash contaminated clothing before reuse.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P330-Rinse mouth.