missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORP buffer +271mV, Hamilton™
Click to view available options
Quantity:
500 mL
Specifications
Physical Form | Liquid |
Quantity | 500 mL |
Solution Type | Buffer |
Solubility Information | Water Soluble |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'